Hydrochloric acid, 37%, for analysis, (max. 0.000001% Hg)
Hydrochloric acid, 37%, for analysis, (max. 0.000001% Hg)
Hydrochloric acid, 37%, for analysis, (max. 0.000001% Hg)
Hydrochloric acid, 37%, for analysis, (max. 0.000001% Hg)
Thermo Scientific Chemicals
Hydrochloric acid, 37%, for analysis, (max. 0.000001% Hg)
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4505500252.5 LGlass Bottle
Catalog number ACR450550025
Price (MYR)
559.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
559.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydrochloric Acid
CAS7647-01-0
Melting Point-35.0°C
Boiling Point57.0°C
Density1.1800g/mL