1-Acetylpyrene, 97%
1-Acetylpyrene, 97%
1-Acetylpyrene, 97%
Thermo Scientific Chemicals
1-Acetylpyrene, 97%
CAS: 3264-21-9 | C18H12O | 244.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4493000505 gGlass bottle
Catalog number ACR449300050
Price (MYR)
1,129.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,129.00
EA
Add to cart
Specifications
Chemical Name or Material1-Acetylpyrene
CAS3264-21-9
Melting Point86.0°C to 89.0°C
Assay Percent Range96% min. (HPLC)
PackagingGlass bottle