L-Lactide, 98%, Thermo Scientific Chemicals
L-Lactide, 98%, Thermo Scientific Chemicals
L-Lactide, 98%, Thermo Scientific Chemicals
L-Lactide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L-Lactide, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ACR44921025025 g
Catalog number ACR449210250
Price (MYR)
844.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
844.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-Lactide
Melting Point95°C to 98°C
CAS4511-42-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement