(-)-Epigallocatechin, 95%, from green tea
(-)-Epigallocatechin, 95%, from green tea
(-)-Epigallocatechin, 95%, from green tea
(-)-Epigallocatechin, 95%, from green tea
Thermo Scientific Chemicals
(-)-Epigallocatechin, 95%, from green tea
CAS: 970-74-1 | C15H14O7 | 306.27 g/mol
Have Questions?
Catalog NumberQuantity
ACR44898010010 mg
Catalog number ACR448980100
Price (MYR)
1,904.00
EA
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (MYR)
1,904.00
EA
Add to cart
Specifications
Chemical Name or Material(-)-Epigallocatechin
CAS970-74-1
Assay Percent Range94% min. (HPLC)
Molecular FormulaC15H14O7
Quantity10 mg