Specifications
Chemical Name or MaterialBenzyl alcohol
CAS100-51-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/phys
View more