Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Thermo Scientific Chemicals
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
CAS: 7758-11-4 | HK2O4P | 174.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR447362500250 gPlastic bottle
Catalog number ACR447362500
Price (MYR)
1,798.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,798.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium phosphate, dibasic
Melting Point340.0°C
CAS7758-11-4
Quantity250 g
EINECS Number231-834-5
View more
For HPLC
RUO – Research Use Only