Mercury(I) nitrate dihydrate, ACS reagent, 9th edition
Mercury(I) nitrate dihydrate, ACS reagent, 9th edition
Mercury(I) nitrate dihydrate, ACS reagent, 9th edition
Mercury(I) nitrate dihydrate, ACS reagent, 9th edition
Thermo Scientific Chemicals
Mercury(I) nitrate dihydrate, ACS reagent, 9th edition
CAS: 7782-86-7 | Hg2N2O6·2H2O | 561.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR447182500250 gPlastic bottle
Catalog number ACR447182500
Price (MYR)
1,374.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,374.00
EA
Add to cart
Specifications
Chemical Name or MaterialMercury(I) nitrate dihydrate
Melting Point70.0°C
CAS7782-86-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.