Acetyl chloride, 99%, ACS reagent, Thermo Scientific™
Acetyl chloride, 99%, ACS reagent, Thermo Scientific™
Acetyl chloride, 99%, ACS reagent, Thermo Scientific™
Acetyl chloride, 99%, ACS reagent, Thermo Scientific™
Acetyl chloride, 99%, ACS reagent, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR447171000100 mLGlass bottle
Catalog number ACR447171000
Price (MYR)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetyl chloride
CAS75-36-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Reacts violently with water.
Corrosive to the respiratory tract.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
View more
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.