Carbon disulfide, 99.9%, ACS reagent
Carbon disulfide, 99.9%, ACS reagent
Carbon disulfide, 99.9%, ACS reagent
Carbon disulfide, 99.9%, ACS reagent
Thermo Scientific Chemicals
Carbon disulfide, 99.9%, ACS reagent
CAS: 75-15- | CS2 | 76.13 g/mol
Have Questions?
Catalog NumberChemical Name or Material
ACR447035000Carbon disulfide
Catalog number ACR447035000
Price (MYR)
645.00
EA
Add to cart
Chemical Name or Material:
Carbon disulfide
Request bulk or custom format
Price (MYR)
645.00
EA
Add to cart
Specifications
Chemical Name or MaterialCarbon disulfide
Melting Point-111.0°C
CAS75-15-
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Highly fla