2-Bromoethyl ether, 90%
2-Bromoethyl ether, 90%
2-Bromoethyl ether, 90%
2-Bromoethyl ether, 90%
Thermo Scientific Chemicals
2-Bromoethyl ether, 90%
CAS: 5414-19-7 | C4H8Br2O | 231.92 g/mol
Have Questions?
Catalog NumberQuantity
ACR44586025025 mL
Catalog number ACR445860250
Price (MYR)
2,267.00
EA
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (MYR)
2,267.00
EA
Add to cart
Specifications
Chemical Name or Material2-Bromoethyl ether
CAS5414-19-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.