1,2-Dichlorobenzene, 99+%, for HPLC
1,2-Dichlorobenzene, 99+%, for HPLC
1,2-Dichlorobenzene, 99+%, for HPLC
1,2-Dichlorobenzene, 99+%, for HPLC
Thermo Scientific Chemicals
1,2-Dichlorobenzene, 99+%, for HPLC
CAS: 95-50-1 | C6H4Cl2 | 147 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR445641000100 mLGlass bottle
Catalog number ACR445641000
Price (MYR)
467.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
467.00
EA
Add to cart
Specifications
Chemical Name or Material1, 2-Dichlorobenzene
CAS95-50-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Harmful if inhaled.
May cause
Health Hazard 3GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritatio