Diclofenac sodium, CAS # 15307-79-6, is a nonsteroidal anti-inflammatory drug (NSAID) that inhibits prostaglandin synthesis.
Have Questions?
Catalog Number
Quantity
Packaging
ACR445250100
10 g
Glass bottle
Catalog number ACR445250100
Price (MYR)
787.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
787.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiclofenac sodium
CAS15307-79-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Toxic if swallowed.
Health Hazard 3GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
View more
General Description
Diclofenac is a is a nonsteroidal anti-inflammatory drug (NSAID) with anti-inflammatory and analgesic properties
Diclofenac has been shown to have anti-tumor capabilities through inhibition of MYC expression and glucose metabolism in models of melanoma and leukemia
Applications
Diclofenac sodium can be used as an antioxidant and free radical scavenger
It has been shown to have anti-bacterial activity against Listeria, E. coli, and M. tuberculosis partially through inhibition of bacterial DNA synthesis
It can be used for the study of metabolic activity as a CYP2C9 substrate
RUO – Research Use Only
General References:
Altman, R; Bosch, B.; Brune, K.; Patrignani, P.; Young, C. Advances in NSAID development: evolution of diclofenac products using pharmaceutical technology. Drugs. 2015, 75(8), 859-77.
Tang, Y.Z.; Liu, Z.Q. Evaluation of the free-radical-scavenging activity of diclofenac acid on the free-radical-induced haemolysis of human erythrocytes. J Pharm Pharmacol. 2006, 58(5), 625-31.
Mazumdar K, Dastidar SG, Park JH, Dutta NK. The anti-inflammatory non-antibiotic helper compound diclofenac: an antibacterial drug target. Eur J Clin Microbiol Infect Dis. 2009, 28(8), 881-91.
Gottfried, E.; Lang, S.A.; Renner, K.; Bosserhoff, A.; Gronwald, W.; Rehli, M.; Einhell, S.; Gedig, I.; Singer, K.; Seilbeck, A.; Mackensen, A.; Grauer, O.; Hau, P.; Dettmer, K.; Andreesen, R.; Oefner, P.J.; Kreutz, M. New aspects of an old drug--diclofenac targets MYC and glucose metabolism in tumor cells. PLoS One. 2013, 8(7), e66987.
Cantoni, L.; Valaperta, R.; Ponsoda, X.; Castell, J.V.; Barelli, D., Rizzardini, M.; Mangolini, A.; Hauri, L.; Villa, P. Induction of hepatic heme oxygenase-1 by diclofenac in rodents: role of oxidative stress and cytochrome P-450 activity. J Hepatol. 2003, 38(6), 776-83.