6-chloronicotinoyl chloride, 97%, Thermo Scientific™
6-chloronicotinoyl chloride, 97%, Thermo Scientific™
6-chloronicotinoyl chloride, 97%, Thermo Scientific™
6-chloronicotinoyl chloride, 97%, Thermo Scientific™
6-chloronicotinoyl chloride, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR44225025025 gGlass bottle
Catalog number ACR442250250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material6-chloronicotinoyl chloride
CAS58757-38-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0