4-Methoxyphenethylmagnesium chloride, 0.5M solution in THF, AcroSeal™
4-Methoxyphenethylmagnesium chloride, 0.5M solution in THF, AcroSeal™
4-Methoxyphenethylmagnesium chloride, 0.5M solution in THF, AcroSeal™
4-Methoxyphenethylmagnesium chloride, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals
4-Methoxyphenethylmagnesium chloride, 0.5M solution in THF, AcroSeal™
CAS: 211115-05-8 | C9H11ClMgO | 194.94 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR44080050050 mLAcroSeal™ Glass Bottle
Catalog number ACR440800500
Price (MYR)
1,292.00
EA
Add to cart
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
1,292.00
EA
Add to cart
Specifications
Chemical Name or Material4-Methoxyphenethylmagnesium chloride
CAS211115-05-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.
May form explosive peroxides.
May cause respiratory irritation.
CAS109-99-9