4-chloro-2,5-dimethoxyaniline, 97%, Thermo Scientific™
4-chloro-2,5-dimethoxyaniline, 97%, Thermo Scientific™
4-chloro-2,5-dimethoxyaniline, 97%, Thermo Scientific™
4-chloro-2,5-dimethoxyaniline, 97%, Thermo Scientific™
Thermo Scientific Chemicals
4-chloro-2,5-dimethoxyaniline, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR437861000100 gGlass bottle
Catalog number ACR437861000
Price (MYR)
705.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
705.00
EA
Add to cart
Specifications
Chemical Name or Material4-chloro-2, 5-dimethoxyaniline
CAS6358-64-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.