Potassium hydroxide, 99.98%, (trace metal basis), pellets
Potassium hydroxide, 99.98%, (trace metal basis), pellets
Potassium hydroxide, 99.98%, (trace metal basis), pellets
Potassium hydroxide, 99.98%, (trace metal basis), pellets
Thermo Scientific Chemicals
Potassium hydroxide, 99.98%, (trace metal basis), pellets
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR437131000100 gPlastic bottle
Catalog number ACR437131000
Price (MYR)
1,007.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,007.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium hydroxide
ColorWhite
Boiling Point1320.0°C
CAS1310-58-3
Health Hazard 1GHS Signal Word: Danger
View more
For ultra trace analysis
RUO – Research Use Only