Aluminum, 99.9995%, (trace metal basis), shot
Aluminum, 99.9995%, (trace metal basis), shot
Aluminum, 99.9995%, (trace metal basis), shot
Aluminum, 99.9995%, (trace metal basis), shot
Thermo Scientific Chemicals
Aluminum, 99.9995%, (trace metal basis), shot
CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR43707025025 gGlass bottle
Catalog number ACR437070250
Price (MYR)
1,326.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,326.00
EA
Add to cart
Specifications
Chemical Name or MaterialAluminum
ColorGray
CAS7429-90-5
Melting Point660.0°C
Boiling Point2327.0°C
View more
For ultra trace analysis
RUO – Research Use Only