Copper, 99.9998%, (trace metal basis), Thermo Scientific™
Copper, 99.9998%, (trace metal basis), Thermo Scientific™
Copper, 99.9998%, (trace metal basis), Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR437051250125 gGlass bottle
Catalog number ACR437051250
Price (MYR)
-
Quantity:
125 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper
ColorBrown-Rust
CAS7440-50-8
Melting Point1083.0°C
Boiling Point2595.0°C
View more
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For ultra trace analysis