Trifluoroacetic acid, 99+%, for spectroscopy
Trifluoroacetic acid, 99+%, for spectroscopy
Trifluoroacetic acid, 99+%, for spectroscopy
Trifluoroacetic acid, 99+%, for spectroscopy
Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, for spectroscopy
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR434161000100 mLGlass bottle
Catalog number ACR434161000
Price (MYR)
1,553.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,553.00
EA
Add to cart
Specifications
Absorbance0.03 max. at 300 nm, 0.03 max. at 320 nm, 0.04 max. at 290 nm, 0.05 max. at 280 nm, 0.1 max. at 270nm, 1 max. at 260 nm
Chemical Name or MaterialTrifluoroacetic acid
Name NoteFor Spectroscopy
CAS76-05-1
Melting Point-15.0°C
View more
For UV/VIS spectroscopy
RUO – Research Use Only