Guanidine thiocyanate, 99+%, for molecular biology
Guanidine thiocyanate, 99+%, for molecular biology
Guanidine thiocyanate, 99+%, for molecular biology
Guanidine thiocyanate, 99+%, for molecular biology
Thermo Scientific Chemicals
Guanidine thiocyanate, 99+%, for molecular biology
Have Questions?
Catalog NumberQuantity
ACR433941000100 g
Catalog number ACR433941000
Price (MYR)
878.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
878.00
EA
Add to cart
Specifications
Chemical Name or MaterialGuanidine thiocyanate
FormCrystalline Powder
Quantity100 g
pHpH 5.0 to 7.0
Purity or Quality Grade≥99.0%, Molecular Biology
View more
Applications
Guanidine thiocyanate is a useful tool for RNA isolation and protein solubilization. It is involved in the quantification of mRNA from hepatocytes. It acts as a medium for blood smaple lystae in order to maintain the integrity of nucleic acid. It is a chaotropic agent, mainly involved in the extraction of DNA and RNA. It is utilized to deactivate the influenza virus.
Solubility
Soluble in water and ethanol
Notes
Light sensitive. Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only