3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
Thermo Scientific Chemicals
3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D
CAS: 24493-21-8 | C6H9D4NaO2Si | 172.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4321200101 gGlass bottle
Catalog number ACR432120010
Price (MYR)
1,388.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,388.00
EA
Add to cart
Specifications
Chemical Name or Material3-(Trimethylsilyl)propionic acid-d4 sodium salt
CAS24493-21-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
For NMR
RUO – Research Use Only