Sulfaguanidine, 98%, Thermo Scientific Chemicals
Sulfaguanidine, 98%, Thermo Scientific Chemicals
Sulfaguanidine, 98%, Thermo Scientific Chemicals
Sulfaguanidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfaguanidine, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR43130025025 gGlass bottle
Catalog number ACR431300250
Price (MYR)
459.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
459.00
EA
Add to cart
Specifications
Chemical Name or MaterialSulfaguanidine
Melting Point190.5°C to 192.0°C
CAS57-67-0
ColorWhite
Health Hazard 1GHS Signal Word: Warning