Anisole, 99%, Extra Dry, AcroSeal™
Anisole, 99%, Extra Dry, AcroSeal™
Anisole, 99%, Extra Dry, AcroSeal™
Anisole, 99%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Anisole, 99%, Extra Dry, AcroSeal™
CAS: 100-66-3 | C7H8O | 108.14 g/mol
Have Questions?
Catalog NumberQuantity
ACR429251000100 mL
Catalog number ACR429251000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAnisole
CAS100-66-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ven