(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, AcroSeal™
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, AcroSeal™
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, AcroSeal™
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, AcroSeal™
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, AcroSeal™
Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, AcroSeal™
CAS: 18107-18-1 | C4H10N2Si | 114.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR429211000100 mLAcroSeal™ Glass Bottle
Catalog number ACR429211000
Price (MYR)
3,759.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
3,759.00
EA
Add to cart
Specifications
Chemical Name or Material(Trimethylsilyl)diazomethane
CAS18107-18-1
CAS Min %32.15
Health Hazard 1GHS Signal Word: Danger
CAS Max %37.23
View more
Alkylation reagent
RUO – Research Use Only