N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
Thermo Scientific Chemicals
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
CAS: 538-75-0 | C13H22N2 | 206.33 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR429021000100 mLAcroSeal™ Glass Bottle
Catalog number ACR429021000
Price (MYR)
508.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
508.00
EA
Add to cart
Chemical Identifiers
CAS538-75-0, 75-09-2
Specifications
Chemical Name or MaterialN, N'-Dicyclohexylcarbodiimide
CAS538-75-0
CAS Min %15.81
Health Hazard 1GHS Signal Word: Danger
CAS Max %18.3
View more
Coupling reagent
RUO – Research Use Only