N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride, 98%
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride, 98%
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride, 98%
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride, 98%
Thermo Scientific Chemicals
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride, 98%
CAS: 21653-40-7 | C19H22N6O4·HCl | 434.88 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4286400101 gGlass bottle
Catalog number ACR428640010
Price (MYR)
5,217.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,217.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride
Melting Point223.0°C
CAS21653-40-7
CAS Min %97.5
CAS Max %100.0