Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™
Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™
Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™
Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™
Thermo Scientific Chemicals
Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™
CAS: 593-04-4 | C8H18BF3O | 198.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR428141000100 mLAcroSeal™ Glass Bottle
Catalog number ACR428141000
Price (MYR)
934.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
934.00
EA
Add to cart
Specifications
Chemical Name or MaterialBoron trifluoride dibutyl etherate
CAS593-04-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
Lewis acid
RUO – Research Use Only