Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
Thermo Scientific Chemicals
Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
CAS: 811-98-3 | CH4O | 36.066 g/mol
Have Questions?
Catalog NumberQuantity
ACR426951000100 mL
Catalog number ACR426951000
Price (MYR)
4,101.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
4,101.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethanol-d4
Density0.8880g/mL
Deuterium (D)99.75% min.
Flash Point12°C
Physical FormLiquid
View more
For NMR
RUO – Research Use Only