Methyl sulfoxide-d6, for NMR, 99.9 atom% D, AcroSeal™
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ACR426941000
100 mL
AcroSeal™ Glass Bottle
Catalog number ACR426941000
Price (MYR)
2,733.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
2,733.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide-d6
Name NoteFor NMR, 99.9 atom% D
CAS2206-27-1
Melting Point20.2°C