Check Order Status
4-chloro-3-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™
Element Search
Structure Search
4-chloro-3-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
Packaging
ACR425840010
1 g
Glass bottle
Catalog number
ACR425840010
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
4-chloro-3-(trifluoromethyl)benzaldehyde
CAS
34328-46-6
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0