2,2,4-Trimethylpentane, 99+%, ACS reagent
2,2,4-Trimethylpentane, 99+%, ACS reagent
2,2,4-Trimethylpentane, 99+%, ACS reagent
2,2,4-Trimethylpentane, 99+%, ACS reagent
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99+%, ACS reagent
CAS: 540-84-1 | C8H18 | 114.232 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4245600252.5 LGlass Bottle
Catalog number ACR424560025
Price (MYR)
898.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
898.00
EA
Add to cart
Specifications
Chemical Name or Material2, 2, 4-Trimethylpentane
Melting Point-107.0°C
Name NoteFor analysis ACS
CAS540-84-1
CAS Min %99.0