Sodium fluoride, ≥99%, ACS reagent, powder, Thermo Scientific Chemicals
Sodium fluoride, ≥99%, ACS reagent, powder, Thermo Scientific Chemicals
Sodium fluoride, ≥99%, ACS reagent, powder, Thermo Scientific Chemicals
Sodium fluoride, ≥99%, ACS reagent, powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium fluoride, ≥99%, ACS reagent, powder, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR4243200505 gPlastic bottle
Catalog number ACR424320050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Boiling Point1700.0°C
Chemical Name or MaterialSodium fluoride
Melting Point993.0°C
CAS7681-49-4
ColorWhite
View more
C-X bond formation reagent
RUO – Research Use Only