Potassium nitrate, 99+%, ACS reagent
Potassium nitrate, 99+%, ACS reagent
Potassium nitrate, 99+%, ACS reagent
Potassium nitrate, 99+%, ACS reagent
Thermo Scientific Chemicals
Potassium nitrate, 99+%, ACS reagent
CAS: 7757-79-1 | KNO3 | 101.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR424151000
Price (MYR)
497.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
497.00
EA
Add to cart
Specifications
Boiling Point400.0°C
Chemical Name or MaterialPotassium nitrate
Melting Point334.0°C
Name Notecrystals
CAS7757-79-1