Potassium hydroxide, ca. 85%, ACS reagent, pellets
Potassium hydroxide, ca. 85%, ACS reagent, pellets
Potassium hydroxide, ca. 85%, ACS reagent, pellets
Potassium hydroxide, ca. 85%, ACS reagent, pellets
Thermo Scientific Chemicals
Potassium hydroxide, ca. 85%, ACS reagent, pellets
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantity
ACR424145000500 g
Catalog number ACR424145000
Price (MYR)
495.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
495.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium Hydroxide
ColorWhite
Name Note85%
Boiling Point1320.0°C
CAS1310-58-3