Potassium Chloride, 99+%, ACS Reagent
Potassium Chloride, 99+%, ACS Reagent
Potassium Chloride, 99+%, ACS Reagent
Potassium Chloride, 99+%, ACS Reagent
Thermo Scientific Chemicals
Potassium Chloride, 99+%, ACS Reagent
CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
25 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number ACR424090050
Price (MYR)
2,041.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
2,041.00
EA
Add to cart
Specifications
Boiling Point1420.0°C
Chemical Name or MaterialPotassium chloride
Melting Point770.0°C
CAS7447-40-7