Acetone, 99.6%, ACS reagent
Acetone, 99.6%, ACS reagent
Acetone, 99.6%, ACS reagent
Acetone, 99.6%, ACS reagent
Thermo Scientific Chemicals
Acetone, 99.6%, ACS reagent
CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4232400252.5 LGlass Bottle
Catalog number ACR423240025
Price (MYR)
732.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
732.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetone
Melting Point-95.0°C
CAS67-64-1
CAS Min %99.5
Health Hazard 1Danger