p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent
CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR421211000100 gPlastic bottle
Catalog number ACR421211000
Price (MYR)
397.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
397.00
EA
Add to cart
Specifications
Chemical Name or Materialp-Toluenesulfonic acid monohydrate
CAS6192-52-5
Health Hazard 1Danger
ColorWhite
Health Hazard 2GHS H Statement: