Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
Thermo Scientific Chemicals
Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
CAS: 5137-46-2 | C12H10Na | 177.202 g/mol
Have Questions?
Catalog NumberQuantity
ACR419423000300 mL
Catalog number ACR419423000
Price (MYR)
1,839.00
BOT
Add to cart
Quantity:
300 mL
Request bulk or custom format
Price (MYR)
1,839.00
BOT
Add to cart
Chemical Identifiers
CAS5137-46-2, 112-36-7
Specifications
Chemical Name or MaterialSodium biphenyl, 20%w/w solution in diethylene glycol diethyl ether
ColorBlack-Blue to Green
Name Note20%w/w Solution in Diethylene Glycol Diethyl Ether
CAS5137-46-2
Health Hazard 1Danger
View more
Reduction reagent
RUO – Research Use Only