Nitrobenzene, 99+%, ACS reagent
Nitrobenzene, 99+%, ACS reagent
Nitrobenzene, 99+%, ACS reagent
Nitrobenzene, 99+%, ACS reagent
Thermo Scientific Chemicals
Nitrobenzene, 99+%, ACS reagent
CAS: 98-95-3 | C6H5NO2 | 123.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
Packaging:
Glass bottle
Catalog number ACR415785000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialNitrobenzene
CAS98-95-3
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0