Nicotine ditartrate dihydrate, 98%
Nicotine ditartrate dihydrate, 98%
Nicotine ditartrate dihydrate, 98%
Nicotine ditartrate dihydrate, 98%
Nicotine ditartrate dihydrate, 98%
Thermo Scientific Chemicals
Nicotine ditartrate dihydrate, 98%
CAS: 6019-06-3 | C18H30N2O14 | 498.44 g/mol
Have Questions?
Catalog NumberQuantity
ACR41566050050 g
Catalog number ACR415660500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNicotine ditartrate dihydrate
Name Note98%
CAS6019-06-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger