Menthofuran, 95%
Menthofuran, 95%
Menthofuran, 95%
Thermo Scientific Chemicals
Menthofuran, 95%
CAS: 494-90-6 | C10H14O | 150.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR41356010010 gGlass bottle
Catalog number ACR413560100
Price (MYR)
702.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
702.00
EA
Add to cart
Specifications
Chemical Name or MaterialMenthofuran
CAS494-90-6
Health Hazard 1GHS Signal Word: Warning
ColorYellow
Density0.9700g/mL