Lanolin, anhydrous, USP
Lanolin, anhydrous, USP
Lanolin, anhydrous, USP
Lanolin, anhydrous, USP
Thermo Scientific Chemicals
Lanolin, anhydrous, USP
CAS: 8006-54-0
Have Questions?
Catalog NumberQuantityPackaging
ACR413005000500 gGlass bottle
Catalog number ACR413005000
Price (MYR)
679.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
679.00
EA
Add to cart
Specifications
Chemical Name or MaterialLanolin
AlkalinityPasses Test
Acidity2.0 ml 0.10 N NaOH max.
CAS8006-54-0
Melting Point38°C to 44°C