2-Methyl-1-propanol, for HPLC
2-Methyl-1-propanol, for HPLC
2-Methyl-1-propanol, for HPLC
2-Methyl-1-propanol, for HPLC
Thermo Scientific Chemicals
2-Methyl-1-propanol, for HPLC
CAS: 78-83-1 | C4H10O | 74.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4126400101 LGlass bottle
Catalog number ACR412640010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Methyl-1-propanol
Name NoteFor HPLC
CAS78-83-1
Absorbance0.01 max. at 330nm, 0.01 max. at 400nm, 0.03 max. at 260nm, 0.05 max. at 250nm, 0.1 max. at 245nm, 0.5 max. at 230nm, 1 max. at 219nm
Health Hazard 1GHS Signal Word: Danger
View more
For HPLC
RUO – Research Use Only