Iodine monochloride, ACS reagent
Iodine monochloride, ACS reagent
Iodine monochloride, ACS reagent
Iodine monochloride, ACS reagent
Thermo Scientific Chemicals
Iodine monochloride, ACS reagent
CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR412361000100 gGlass bottle
Catalog number ACR412361000
Price (MYR)
1,028.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,028.00
EA
Add to cart
Specifications
Boiling Point97.0°C
Chemical Name or MaterialIodine monochloride
Melting Point26.0°C
CAS7790-99-0
ColorBrown-Red
View more
C-X bond formation reagent
RUO – Research Use Only