Heparin sodium
Heparin sodium
Heparin sodium
Thermo Scientific Chemicals
Heparin sodium
Heparin Sodium Salt, CAS # 9041-08-1, is derived from porcine intestinal mucosa and shows blood anti-coagulant activity. | CAS: 9041-08-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
View all
Glass bottle
Catalog number ACR411212500
Price (MYR)
789.00
EA
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
789.00
EA
Add to cart
Specifications
Chemical Name or MaterialHeparin sodium
CAS9041-08-1
Absorbance≤0.2% as nucleotides, 0.2 max. (0.4% in water) at 260nm, Protein and nucleotide impurities:, Protein and nucleotide impurities: at 0nm
Health Hazard 1HYGROSCOPIC
ColorWhite
View more
General Description
  • Heparin sodium salt displays anti-coagulant activity. It can also efficiently inhibit mesenchymal cell proliferation and promotes the proliferation of primary epithelial cells
  • This compound can bind to antithrombin III and inhibit coagulation proteases
Applications
  • In in vitro studies, it is used as the main component of serum-free media to prevent cell aggregation
  • Heparin sodium salt can also induce the growth of human embryonic stem cells in a defined serum-free medium, acting as a cofactor for a basic fibroblast growth factor (FGF-2)
  • It is also applied in laboratory studies related to stem cell differentiation and growth
  • In in vitro studies this compound exhibits cytotoxic action inducing cancer cell death by interfering with transcription factor activity
RUO – Research Use Only