Diethyl phosphate, 96%
Diethyl phosphate, 96%
Diethyl phosphate, 96%
Diethyl phosphate, 96%
Thermo Scientific Chemicals
Diethyl phosphate, 96%
CAS: 598-02-7 | C4H11O4P | 154.1 g/mol
Have Questions?
Catalog NumberQuantity
ACR4074200505 g
Catalog number ACR407420050
Price (MYR)
1,119.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,119.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiethyl phosphate
CAS598-02-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
CAS7664-38-2