Cholic acid, sodium salt, 98+%
Cholic acid, sodium salt, 98+%
Cholic acid, sodium salt, 98+%
Cholic acid, sodium salt, 98+%
Thermo Scientific Chemicals
Cholic acid, sodium salt, 98+%
Cholic acid, sodium salt, CAS # 361-09-1, is a bile acid that can be non-denaturing detergent and can be used for membrane protein extraction. | CAS: 361-09-1 | C24H39NaO5 | 430.56 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR405191000100 gGlass bottle
Catalog number ACR405191000
Price (MYR)
887.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
887.00
EA
Add to cart
Specifications
Chemical Name or MaterialCholic acid, sodium salt
CAS361-09-1
ColorWhite
pH7.5 to 9.5 (5% aq. soln.)
Physical FormPowder
View more
General description
• Cholic acid, sodium salt, is a bile acid that can aid digestion of fats and certain nutrients
Application
• It can facilitate digestion of lipids
• It is a non-denaturing detergent
• It can be used in combination with urea for extraction of membrane protein and protein complexes
RUO – Research Use Only
General References:
  1. Hori, H., et al. Isolation and characterization of two 20S proteasomes from the endoplasmic reticulum of rat liver microsomes. J Biochem 1999, 126(4), 722-730.
  2. Helenius, A; McCaslin, D. R.; Fries, E.; Tanford, C. [63] Properties of detergents. Methods Enzymol 1979, 56, 734-749.
  3. Hjelmeland LM, Chrambach A. Solubilization of functional membrane proteins. Methods Enzymol 1984, 104, 305-18.