Bromothymol Blue sodium salt, ACS reagent
Bromothymol Blue sodium salt, ACS reagent
Bromothymol Blue sodium salt, ACS reagent
Bromothymol Blue sodium salt, ACS reagent
Thermo Scientific Chemicals
Bromothymol Blue sodium salt, ACS reagent
CAS: 34722-90-2 | C27H27Br2NaO5S | 646.37 g/mol
Have Questions?
Catalog NumberQuantity
ACR40326010010 g
Catalog number ACR403260100
Price (MYR)
538.00
BOT
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
538.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBromothymol Blue sodium salt
CAS34722-90-2
ClarityPasses Test
Molecular FormulaC27H27Br2NaO5S
MDL NumberMFCD00077263,MFCD00077263,MFCD00077263
View more
Indicator
RUO – Research Use Only