Aniline Blue, sodium salt, pure, water soluble, certified
Aniline Blue, sodium salt, pure, water soluble, certified
Aniline Blue, sodium salt, pure, water soluble, certified
Aniline Blue, sodium salt, pure, water soluble, certified
Thermo Scientific Chemicals
Aniline Blue, sodium salt, pure, water soluble, certified
CAS: 28983-56-4 | C37H26N3Na2O9S3 | 798.79 g/mol
Have Questions?
Catalog NumberQuantity
ACR40118025025 g
Catalog number ACR401180250
Price (MYR)
627.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
627.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAniline Blue, sodium salt
CAS28983-56-4
Health Hazard 1Signal Word: WARNING!
Absorption Ratio0.96 to 1.09
Health Hazard 2GHS H Statement: