View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetone
Melting Point-95.0°C
Name NoteReagent ACS, Spectro Grade
Absorbance0.01 max. at 400nm, 0.02 max. at 350nm, 0.1 max. at 340nm, 1 max. at 330nm
CAS67-64-1